Learn More
3M™ PELTOR™ LiteCom™ Plus Headset
3M™ PELTOR™ LiteCom™ Plus Headset is based on a robust basic design with a number of added features.
Brand: 3M™ 7000108543
Code : NEW
Additional Details : Weight : 1.09800kg Transport : UN number : 3481 Chem class : 9
Description
- Headset with built in communication radio allows two-way conversation up to 3000 meters
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- It's easy to connect a mobile phone or external communication radio, regardless of the frequency
- Microphone software is programmed only to pick up speech and effectively filters out all other sounds
- 121 sub-channels allow several users to communicate individually using the same channel
- Communication on 8 channels in the PMR 446MHz band
- Up to 1 kilometer range
Specifications
Communication Headset | |
33 dB | |
CE Approved |