Learn More
3M™ PELTOR™ LiteCom™ Plus Headset
3M™ PELTOR™ LiteCom™ Plus Headset is based on a robust basic design with a number of added features.
£704.00 - £970.00
Specifications
Product Type | Communication Headset |
---|---|
Certifications/Compliance | CE Approved |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
15779997
|
3M™
7000108543 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||||
15789997
|
3M™
7000108544 |
Helmet Mounted |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15799997
|
3M™
7000108545 |
Neckband |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Headset with built in communication radio allows two-way conversation up to 3000 meters
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- It's easy to connect a mobile phone or external communication radio, regardless of the frequency
- Microphone software is programmed only to pick up speech and effectively filters out all other sounds
- 121 sub-channels allow several users to communicate individually using the same channel
- Communication on 8 channels in the PMR 446MHz band
- Up to 1 kilometer range
Specifications
Communication Headset | |
CE Approved |