missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEA-15 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
£279.00 - £401.00
Specifications
| Antigen | PEA-15 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605268
|
Novus Biologicals
NBP3-05544-100ul |
100 μg |
£401.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600358
|
Novus Biologicals
NBP3-05544-50ul |
50 μg |
£279.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PEA-15 Monoclonal antibody specifically detects PEA-15 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PEA-15 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| HMAT115kD, PEA-15, PED15 kDa phosphoprotein enriched in astrocytes, phosphoprotein enriched in astrocytes 15, Phosphoprotein enriched in diabetes | |
| Full length human PEA-15 as expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, Cellular Markers | |
| 50% PBS, 50% glycerol | |
| 8682 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title