missing translation for 'onlineSavingsMsg'
Learn More

PEA-15 Antibody, Novus Biologicals™

Product Code. 18605268 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18605268 100 μg 100µL
18600358 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18605268 Supplier Novus Biologicals Supplier No. NBP305544100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEA-15 Monoclonal antibody specifically detects PEA-15 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEA-15
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000
Formulation 50% PBS, 50% glycerol
Gene Alias HMAT115kD, PEA-15, PED15 kDa phosphoprotein enriched in astrocytes, phosphoprotein enriched in astrocytes 15, Phosphoprotein enriched in diabetes
Host Species Mouse
Immunogen Full length human PEA-15 as expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 8682
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.