missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20989-1-AP-150UL
This item is not returnable.
View return policy
Description
PDX1 (pancreas duodenum homeobox-1, also known as IPF-1, IDX-1, and STF-1) is a transcription factor that has been shown to activate insulin, glucokinase, IAPP, and somatostatin gene transcription. Significant interest surrounds this proteins glucose-dependent regulation of insulin gene transcription. Additionally, pdx-1 has been studied for its role in pancreatic development which includes maturation and differentiation of pancreatic precursor cells. Mutations in the pdx-1 gene have been shown to cause pancreatic agenesis, maturity-onset diabetes of the young, and type II diabetes.Specifications
| PDX1 | |
| Polyclonal | |
| Unconjugated | |
| PDX1 | |
| Glucose sensitive factor, GSF, IDX 1, Insulin promoter factor 1, Insulin upstream factor 1, IPF 1, IPF1, Islet/duodenum homeobox 1, IUF 1, IUF1, MODY4, PDX 1, PDX1, STF 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18609, 3651 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52945, P52946 | |
| PDX1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction