missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Tekniske data
| Antigen | PDX1 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
16810585
|
Proteintech
20989-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800585
|
Proteintech
20989-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDX1 (pancreas duodenum homeobox-1, also known as IPF-1, IDX-1, and STF-1) is a transcription factor that has been shown to activate insulin, glucokinase, IAPP, and somatostatin gene transcription. Significant interest surrounds this proteins glucose-dependent regulation of insulin gene transcription. Additionally, pdx-1 has been studied for its role in pancreatic development which includes maturation and differentiation of pancreatic precursor cells. Mutations in the pdx-1 gene have been shown to cause pancreatic agenesis, maturity-onset diabetes of the young, and type II diabetes.Specifications
| PDX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P52945, P52946 | |
| 18609, 3651 | |
| Peptide | |
| Primary | |
| -20°C | |
| PDX1 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Glucose sensitive factor, GSF, IDX 1, Insulin promoter factor 1, Insulin upstream factor 1, IPF 1, IPF1, Islet/duodenum homeobox 1, IUF 1, IUF1, MODY4, PDX 1, PDX1, STF 1 | |
| PDX1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title