Learn More
Invitrogen™ PDK4 (PDKH4) Recombinant Human Protein
Inhhibition of PDHKs is viewed as a potential therapeutic treament for type II diabetes.
Brand: Invitrogen™ PR8253A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex.
Specifications
NP_002603.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
100μg | |
RUO - research use only | |
PDK-4 (PDKH4) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
73.9kDa | |
Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PDHK Kinase Family | |
Recombinant | |
Baculovirus |