missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDK4 (PDKH4) Recombinant Human Protein
Inhhibition of PDHKs is viewed as a potential therapeutic treament for type II diabetes.
£1602.00 - £6900.00
Specifications
Accession Number | NP_002603.1 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. |
Molecular Weight (g/mol) | 73.9kDa |
Storage Requirements | Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458663
|
Invitrogen™
PR8253A |
100μg |
£1602.00
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458793
|
Invitrogen™
PV5711 |
1mg |
£6900.00
1mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex.
Specifications
NP_002603.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PDHK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
73.9kDa | |
RUO - research use only | |
PDK-4 (PDKH4) | |
Human | |
GST Tag | |
Frozen |