missing translation for 'onlineSavingsMsg'
Learn More

PDI Rabbit, anti-Human, Unconjugated, Clone: 4K17, Proteintech

Rabbit Recombinant Monoclonal Antibody

£135.00 - £360.00

Specifications

Antigen PDI
Clone 4K17
Concentration 500 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17241546
View Documents
Proteintech
80170-1-RR-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17231546
View Documents
Proteintech
80170-1-RR-100UL
100 μL
£360.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PDI antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PDI. This antibody recognizes human, mouse, rat antigen. The PDI antibody has been validated for the following applications: IF, IHC, WB,ELISA analysis.

Specifications

Specifications

PDI
500 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat
Primary
4K17
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Rabbit
5034
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.