Learn More
PDI Rabbit, anti-Human, Unconjugated, Clone: 4K17, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 80170-1-RR-20UL
Description
The PDI antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PDI. This antibody recognizes human, mouse, rat antigen. The PDI antibody has been validated for the following applications: IF, IHC, WB,ELISA analysis.
Specifications
| PDI | |
| Monoclonal | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 4K17 | |
| Unconjugated | |
| 20 μL | |
| 5034 | |
| Liquid |