missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDHA2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79536
This item is not returnable.
View return policy
Description
PDHA2 Polyclonal specifically detects PDHA2 in Human samples. It is validated for Western Blot.
Specifications
| PDHA2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 1.2.4.1, MGC149517, MGC149518, mitochondrial, PDHAL, PDHE1-A type II, pyruvate dehydrogenase (lipoamide) alpha 2, pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, pyruvate dehydrogenase, E1-alpha polypeptide, testis specific | |
| Rabbit | |
| 43 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 86%; Horse: 85%; Dog: 79%; Pig: 79%; Zebrafish: 79%; Guinea pig: 79%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_005381 | |
| PDHA2 | |
| Synthetic peptide directed towards the N terminal of human PDHA2The immunogen for this antibody is PDHA2. Peptide sequence RMELKADQLYKQKFIRGFCHLCDGQEACCVGLEAGINPSDHVITSYRAHG. | |
| Affinity purified | |
| RUO | |
| 5161 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction