missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE9A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£219.00 - £369.00
Specifications
| Antigen | PDE9A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225310
|
Novus Biologicals
NBP1-00641SS |
0.025 mL |
£219.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776603
|
Novus Biologicals
NBP1-00641 |
0.05 mL |
£369.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE9A Polyclonal specifically detects PDE9A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| PDE9A | |
| Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:400 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A | |
| PDE9A | |
| IgG | |
| Affinity Purified | |
| 68 kDa |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| 5152 | |
| Sequence from the C-terminal region of PDE9A | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title