missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE9A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-00641SS
This item is not returnable.
View return policy
Description
PDE9A Polyclonal specifically detects PDE9A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PDE9A | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| PDE9A | |
| Sequence from the C-terminal region of PDE9A | |
| Affinity Purified | |
| RUO | |
| 5152 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:1000, Immunohistochemistry-Paraffin 1:400 | |
| CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A | |
| Rabbit | |
| 68 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction