missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE8A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£200.00 - £558.00
Specifications
| Antigen | PDE8A |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227804
|
Novus Biologicals
NB300-643-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18213661
|
Novus Biologicals
NB300-643-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149789
|
Novus Biologicals
NB300-643 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE8A Polyclonal specifically detects PDE8A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PDE8A | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| 5151 | |
| Synthetic peptide (Human) (C terminal). | |
| Primary |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse, Rat, Human | |
| PDE8A | |
| IgG | |
| PDE8A |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title