missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE8A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-643
This item is not returnable.
View return policy
Description
PDE8A Polyclonal specifically detects PDE8A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PDE8A | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| PDE8A | |
| Synthetic peptide (Human) (C terminal). | |
| 0.1 mg | |
| Lipid and Metabolism, Signal Transduction | |
| 5151 | |
| Reconstitute with 50 ul of distilled water to a final concentration is 1.0 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PDE8A | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction