missing translation for 'onlineSavingsMsg'
Learn More

PDE6A Mouse, anti-Human, Unconjugated, Clone: 1B3B4, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen PDE6A
Clone 1B3B4
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17271486
View Documents
Proteintech
67832-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17261486
View Documents
Proteintech
67832-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PDE6A(Rod cGMP-specific 3′,5′-cyclic phosphodiesterase subunit alpha) is also named as GMP-PDE alpha, PDE V-B1, PDEA and belongs to the cyclic nucleotide phosphodiesterase family. PDE6A contains an open reading frame capable of coding for a polypeptide of 859 amino acids and about 100 kDa. It is a subunit of a key phototransduction enzyme which participates in processes of transmission and amplification of the visual signal. The expression of PDE6 alpha in retina is essential for normal expression of PDE6 beta and PDE6 gama.

Specifications

Specifications

PDE6A
1000 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat
Primary
1B3B4
Western Blot, ELISA
Unconjugated
Mouse
5145
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.