Learn More
PDE6A Mouse, anti-Human, Unconjugated, Clone: 1B3B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67832-1-IG-150UL
Description
PDE6A(Rod cGMP-specific 3′,5′-cyclic phosphodiesterase subunit alpha) is also named as GMP-PDE alpha, PDE V-B1, PDEA and belongs to the cyclic nucleotide phosphodiesterase family. PDE6A contains an open reading frame capable of coding for a polypeptide of 859 amino acids and about 100 kDa. It is a subunit of a key phototransduction enzyme which participates in processes of transmission and amplification of the visual signal. The expression of PDE6 alpha in retina is essential for normal expression of PDE6 beta and PDE6 gama.
Specifications
| PDE6A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, ELISA | |
| 1B3B4 | |
| Unconjugated | |
| 150 μL | |
| 5145 | |
| Liquid |