missing translation for 'onlineSavingsMsg'
Learn More

PDE2A Antibody (11C6), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen PDE2A
Clone 11C6
Dilution ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications ELISA, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30037041
View Documents
Novus Biologicals
NBP3-26434-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30025416
View Documents
Novus Biologicals
NBP3-26434-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PDE2A Monoclonal antibody specifically detects PDE2A in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications

Specifications

PDE2A
ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated
A synthesized peptide derived from Human PDE2A [UniProt O00408]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
11C6
ELISA, Immunocytochemistry
Unconjugated
Rabbit
Signal Transduction
PBS, pH 7.4, 150mM NaCl and 50% glycerol
5138
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.