missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE2A Antibody (11C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26434-50ul
This item is not returnable.
View return policy
Description
PDE2A Monoclonal antibody specifically detects PDE2A in Human samples. It is validated for ELISA, Immunocytochemistry
Specifications
| PDE2A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry | |
| 11C6 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| cGMP-dependent 3'-5'-cyclic phosphodiesterase, cGMP-stimulated phosphodiesterase 1, cGMP-stimulated phosphodiesterase 4, cGSPDE, CGS-PDE, Cyclic GMP-stimulated phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE2A1, PED2A4, phosphodiesterase 2A, cGMP-stimulated | |
| A synthesized peptide derived from Human PDE2A [UniProt O00408] | |
| 50 μL | |
| Signal Transduction | |
| 5138 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction