missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE10A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£220.00 - £603.00
Specifications
| Antigen | PDE10A |
|---|---|
| Concentration | 0.7 mg/mL |
| Dilution | Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280785
|
Novus Biologicals
NB300-645-0.025MG |
0.025 mg |
£220.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077224
|
Novus Biologicals
NB300-645-0.05MG |
0.05 mg |
£366.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18727273
|
Novus Biologicals
NB300-645 |
0.1 mg |
£603.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDE10A Polyclonal specifically detects PDE10A in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| PDE10A | |
| Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Q9Y233 | |
| PDE10A | |
| IgG | |
| This has no cross-reactivity against any other PDE family members tested so far. |
| 0.7 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, ELISA, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Monkey, Mouse, Rat, Human | |
| 10846 | |
| Synthetic peptide taken within amino acid region 300-350 on human PDE10A2 protein common to all PDE10A variants. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title