missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE10A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB300-645-0.05MG
This item is not returnable.
View return policy
Description
PDE10A Polyclonal specifically detects PDE10A in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PDE10A | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4; cryoprotective agents; HEPES; stabilizing protein (BSA 0.5%), and glycerol (30%). with 0.02% sodium azide | |
| cAMP and cAMP-inhibited cGMP 3′-5′-cyclic phosphodiesterase 10A, dJ416F21.1 (phosphodiesterase 10A), EC 3.1.4, EC 3.1.4.17, EC 3.1.4.35, FLJ11894, FLJ25677, HSPDE10A, phosphodiesterase 10A, phosphodiesterase 10A1 (PDE10A1) | |
| Rabbit | |
| 0.05 mg | |
| 10846 | |
| Human, Mouse, Rat, Monkey | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.7 mg/mL | |
| Western Blot > 1:500, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation > 1:250, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9Y233 | |
| PDE10A | |
| Synthetic peptide taken within amino acid region 300-350 on human PDE10A2 protein common to all PDE10A variants. | |
| Primary | |
| This has no cross-reactivity against any other PDE family members tested so far. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction