missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PDCD4 |
|---|---|
| Concentration | 0.6 mg/mL |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16830374
|
Proteintech
12587-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16820374
|
Proteintech
12587-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.Specifications
| PDCD4 | |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q53EL6, Q61823, Q9JID1 | |
| 18569, 27250, 64031 | |
| PDCD4 Fusion Protein Ag3288 | |
| Primary | |
| -20°C | |
| PDCD4 |
| 0.6 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a | |
| PDCD4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title