missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12587-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.Specifications
| PDCD4 | |
| Polyclonal | |
| Unconjugated | |
| PDCD4 | |
| H731, Nuclear antigen H731 like, PDCD4, Protein 197/15a | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18569, 27250, 64031 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q53EL6, Q61823, Q9JID1 | |
| PDCD4 | |
| PDCD4 Fusion Protein Ag3288 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction