missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L2/B7-DC/PDCD1LG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-88964
This item is not returnable.
View return policy
Description
PD-L2/B7-DC/PDCD1LG2 Polyclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PD-L2/B7-DC/PDCD1LG2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PDCD1LG2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GYPLAEVSWPNVSVPANTSHSRTPEGLYQVTSVLRLKPPPGRNFSCVFWNTHVRELTLASIDLQSQMEPRTHPT | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80380 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction