missing translation for 'onlineSavingsMsg'
Learn More

PD 224378, TRC

Product Code. 30007937
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007937 10 mg 10mg
30005001 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007937 Supplier Toronto Research Chemicals Supplier No. TRCP21746010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 466678-44-4
Molecular Formula C20H35NO11
Molecular Weight (g/mol) 465.49
Synonym (4S)-1-(4-O-β-D-Galactopyranosyl-β-D-glucopyranosyl)-4-(2-methylpropyl)-2-pyrrolidinone, (S)-1-((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)-4-isobutylpyrrolidin-2-one;
IUPAC Name (4S)-1-[(2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]-4-(2-methylpropyl)pyrrolidin-2-one
SMILES O[C@H]1[C@@H](O)[C@@H](CO)O[C@H](O[C@H]2C(O)[C@@H](O)[C@@H](N3C(C[C@H](CC(C)C)C3)=O)O[C@@H]2CO)[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H35NO11/c1-8(2)3-9-4-12(24)21(5-9)19-16(28)15(27)18(11(7-23)30-19)32-20-17(29)14(26)13(25)10(6-22)31-20/h8-11,13-20,22-23,25-29H,3-7H2,1-2H3/t9-,10+,11+,13-,14-,15+,16+,17+,18+,19+,20-/m0/s1
Formula Weight 465.221
Percent Purity >95%
Chemical Name or Material (4S)-Pregabalin Amide Lactose Adduct
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.