missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1/CD279 Mouse anti-Human, Mouse, Rat, Clone: 4H4D1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PD-1/CD279 |
|---|---|
| Clone | 4H4D1 |
| Concentration | 1.21 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867895
|
Proteintech
66220-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857895
|
Proteintech
66220-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.Specifications
| PD-1/CD279 | |
| 1.21 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD279, hPD 1, hPD l, PD1, PD-1, PD-1/CD279, PDCD1, programmed cell death 1, Protein PD 1, SLEB2 | |
| PDCD1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4H4D1 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q02242, Q15116 | |
| 18566, 301626, 5133 | |
| PD-1/CD279 Fusion Protein Ag12470 | |
| Primary | |
| -20°C | |
| PDCD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title