missing translation for 'onlineSavingsMsg'
Learn More

PD-1/CD279 Mouse anti-Human, Mouse, Rat, Clone: 4H4D1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66220-1-IG-150UL

 View more versions of this product

Product Code. 16857895

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.
Specifications

Specifications

PD-1/CD279
Monoclonal
1.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02242, Q15116
PDCD1
PD-1/CD279 Fusion Protein Ag12470
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
4H4D1
Unconjugated
PDCD1
CD279, hPD 1, hPD l, PD1, PD-1, PD-1/CD279, PDCD1, programmed cell death 1, Protein PD 1, SLEB2
Mouse
Protein A
RUO
18566, 301626, 5133
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.