missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCTAIRE3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | PCTAIRE3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619841
|
Novus Biologicals
NBP2-94133-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673252
|
Novus Biologicals
NBP2-94133-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCTAIRE3 Polyclonal antibody specifically detects PCTAIRE3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| PCTAIRE3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 5129 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Cell division protein kinase 18, cyclin-dependent kinase 18, EC 2.7.11, EC 2.7.11.22, PCTAIRE, PCTAIRE3, PCTAIRE-motif protein kinase 3, PCTK3PCTAIRE protein kinase 3, Serine/threonine-protein kinase PCTAIRE-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 373-474 of human CDK18 (NP_002587.2). EFRTYSFPCYLPQPLINHAPRLDTDGIHLLSSLLLYESKSRMSAEAALSHSYFRSLGERVHQLEDTASIFSLKEIQLQKDPGYRGLAFQQPGRGKNRRQSIF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title