missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Small and compact design is stackable, saving valuable space in the freezer
- Lid hinge design allows lid to to sit at 180 degrees
- Transparent lid protects samples and allows for quick assessment of contents
- Embossed numbers and letters on the wells provide easy identification and tracking of samples
- Light-weighted construction made durable for reusability
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Blue |
| Holds | Microtubes (0.2 mL) |
| Material | Polypropylene |
| Positions | 1 |
| Shape | Rectangular |
| Stackable | Yes, When Empty |
| Sterility | Non-sterile |
| For Use With (Equipment) | 0.2 mL tubes both individually and in strips of 8, as well as 32-place assay plates (8 x 4) |
| Length (Metric) | 9.8 cm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?