missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PCM1 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899334
|
Proteintech
19856-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889334
|
Proteintech
19856-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a component of centriolar satellites, which are electron dense granules scattered around centrosomes. Inhibition studies show that this protein is essential for the correct localization of several centrosomal proteins, and for anchoring microtubules to the centrosome. Chromosomal aberrations involving this gene are associated with papillary thyroid carcinomas and a variety of hematological maligcies, including atypical chronic myeloid leukemia and T-cell lymphoma.Specifications
| PCM1 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q15154, Q9R0L6 | |
| 18536, 5108 | |
| PCM1 Fusion Protein Ag13929 | |
| Primary | |
| -20°C | |
| PCM1 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hPCM 1, PCM 1, PCM1, pericentriolar material 1, PTC4 | |
| PCM1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title