missing translation for 'onlineSavingsMsg'
Learn More

PCM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16899334 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16899334 20 μL 20µL
16889334 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16899334 Supplier Proteintech Supplier No. 198561AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The protein encoded by this gene is a component of centriolar satellites, which are electron dense granules scattered around centrosomes. Inhibition studies show that this protein is essential for the correct localization of several centrosomal proteins, and for anchoring microtubules to the centrosome. Chromosomal aberrations involving this gene are associated with papillary thyroid carcinomas and a variety of hematological maligcies, including atypical chronic myeloid leukemia and T-cell lymphoma.

Specifications

Antigen PCM1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PCM1
Gene Accession No. Q15154, Q9R0L6
Gene Alias hPCM 1, PCM 1, PCM1, pericentriolar material 1, PTC4
Gene Symbols PCM1
Host Species Rabbit
Immunogen PCM1 Fusion Protein Ag13929
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18536, 5108
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.