missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDH11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79198
This item is not returnable.
View return policy
Description
PCDH11 Polyclonal specifically detects PCDH11 in Mouse samples. It is validated for Western Blot.
Specifications
| PCDH11 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PCDH11, PCDH11Y, PCDH22, PCDH-PC, PCDHX, PCDH-X, PCDHY, PCDH-Y, protein phosphatase 1, regulatory subunit 119, protocadherin 11X, protocadherin 22, Protocadherin on the X chromosome, Protocadherin on the Y chromosome, Protocadherin prostate cancer, protocadherin-11 X-linked, Protocadherin-11 Y-linked, Protocadherin-22, protocadherin-PC, Protocadherin-S | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27328 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_001074854 | |
| PCDH11 | |
| The immunogen for this antibody is Pcdh11x. Peptide sequence LNQSSMLLIKVKDENDNAPVFTQSFISLSVPENNSPGAQLTKISATDADS. | |
| 100 μL | |
| Primary | |
| The immunogen sequence has 100% identity to both PCDH11X and PCDH11Y. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction