missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCBD1 Recombinant Rabbit Monoclonal Antibody (24GB785)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | PCBD1 |
|---|---|
| Clone | 24GB785 |
| Concentration | 1.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275795
![]() |
Invitrogen™
MA557540 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275107
|
Invitrogen™
MA557541 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCBD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
PCBD1 encodes pterin-4 alpha-carbinolamine dehydratase, an enzyme involved in phenylalanine hydroxylation. A deficiency of this enzyme leads to hyperphenylalaninemia. The enzyme regulates the homodimerization of the transcription factor hepatocyte nuclear factor 1 (HNF1).Specifications
| PCBD1 | |
| 1.5 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61457 | |
| 5092 | |
| A synthesized peptide derived from human PCBD1 (aa 10-30). | |
| Antibody |
| 24GB785 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| PCBD1 | |
| 4-alpha-hydroxy-tetrahydropterin dehydratase; 6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1); DCOH; Dimerization cofactor of hepatocyte nuclear factor 1-alpha; dimerization cofactor of HNF1; dimerizing cofactor for HNF1; Pcbd; PCBD1; PCD; Phenylalanine hydroxylase-stimulating protein; PHS; pt; pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 1; pterin carbinolamine dehydratase; pterin-4 alpha-carbinolamine dehydratase 1; pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha; pterin-4-alpha-carbinolamine dehydratase; TCF1 | |
| PCBD1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title