missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PCBD1 Recombinant Rabbit Monoclonal Antibody (24GB785)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557541

 View more versions of this product

Product Code. 30275107

  • £172.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PCBD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

PCBD1 encodes pterin-4 alpha-carbinolamine dehydratase, an enzyme involved in phenylalanine hydroxylation. A deficiency of this enzyme leads to hyperphenylalaninemia. The enzyme regulates the homodimerization of the transcription factor hepatocyte nuclear factor 1 (HNF1).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCBD1
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P61457
PCBD1
A synthesized peptide derived from human PCBD1 (aa 10-30).
20 μL
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
24GB785
Unconjugated
PCBD1
4-alpha-hydroxy-tetrahydropterin dehydratase; 6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1); DCOH; Dimerization cofactor of hepatocyte nuclear factor 1-alpha; dimerization cofactor of HNF1; dimerizing cofactor for HNF1; Pcbd; PCBD1; PCD; Phenylalanine hydroxylase-stimulating protein; PHS; pt; pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 1; pterin carbinolamine dehydratase; pterin-4 alpha-carbinolamine dehydratase 1; pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha; pterin-4-alpha-carbinolamine dehydratase; TCF1
Rabbit
Affinity chromatography
RUO
5092
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.