missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PBK |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16815614
|
Proteintech
16110-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16805614
|
Proteintech
16110-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.Specifications
| PBK | |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q96KB5, Q9JJ78 | |
| 290326, 52033, 55872 | |
| PBK Fusion Protein Ag9033 | |
| Primary | |
| -20°C | |
| PBK |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cancer/testis antigen 84, CT84, MAPKK like protein kinase, Nori 3, PBK, PDZ binding kinase, SPK, TOPK | |
| Pbk | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title