Learn More
PBK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16110-1-AP-150UL
 View more versions of this product
Product Code. 16805614
  • £349.00 / 150µL
Description
This genes encodes a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
Specifications
Specifications
PBK
Polyclonal
Unconjugated
PBK
Cancer/testis antigen 84, CT84, MAPKK like protein kinase, Nori 3, PBK, PDZ binding kinase, SPK, TOPK
Rabbit
Antigen Affinity Chromatography
RUO
290326, 52033, 55872
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96KB5, Q9JJ78
Pbk
PBK Fusion Protein Ag9033
150 μL
Primary
Human, Mouse, Rat