missing translation for 'onlineSavingsMsg'
Learn More

PAX6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen PAX6
Concentration 0.4 mg/mL
Applications Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16859973
View Documents
Proteintech
12323-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16849973
View Documents
Proteintech
12323-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.
Specifications

Specifications

PAX6
Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence
Unconjugated
Rabbit
Mouse, Human
P26367, P63015
18508, 5080
PAX6 Fusion Protein Ag2984
Primary
-20°C
PAX6
0.4 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
AN, AN2, Aniridia type II protein, D11S812E, MGDA, Oculorhombin, paired box 6, Paired box protein Pax 6, PAX6, WAGR
PAX6
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.