missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12323-1-AP-20UL
This item is not returnable.
View return policy
Description
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Specifications
| PAX6 | |
| Polyclonal | |
| Unconjugated | |
| PAX6 | |
| AN, AN2, Aniridia type II protein, D11S812E, MGDA, Oculorhombin, paired box 6, Paired box protein Pax 6, PAX6, WAGR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18508, 5080 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P26367, P63015 | |
| PAX6 | |
| PAX6 Fusion Protein Ag2984 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction