missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax4 Antibody (3C12), Novus Biologicals™
Mouse Monoclonal Antibody
£187.00 - £416.00
Specifications
| Antigen | Pax4 |
|---|---|
| Clone | 3C12 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18132959
|
Novus Biologicals
NBP2-37644 |
0.1 mL |
£416.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629535
|
Novus Biologicals
NBP2-37644-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax4 Monoclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Pax4 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| 5078 | |
| Purified recombinant fragment of human PAX4 expressed in E. Coli. | |
| Primary | |
| 38 kDa |
| 3C12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
| PAX4 | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title