missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax4 Antibody (3C12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37644
This item is not returnable.
View return policy
Description
Pax4 Monoclonal specifically detects Pax4 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Pax4 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| PAX4 | |
| Purified recombinant fragment of human PAX4 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 5078 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 3C12 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4 | |
| Mouse | |
| 38 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction