missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax2, Mouse anti-Human, Clone: PAX2/1104, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £405.00
Specifications
| Antigen | Pax2 |
|---|---|
| Clone | PAX2/1104 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256147
|
Novus Biologicals
NBP2-79742-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18236557
|
Novus Biologicals
NBP2-79742-100UG |
100 μg |
£405.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax2 Monoclonal specifically detects Pax2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Pax2 | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
| PAX2 | |
| IgG2b κ | |
| Protein A or G purified | |
| This antibody recognizes a protein of 42kDa, which is identified as Pax2. Pax2 has been used as a marker for the identification of renal cell carcinoma and ovarian carcinoma by immunohistochemistry. |
| PAX2/1104 | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q02962, Q02962 | |
| 5076 | |
| Recombinant fragment (around aa 223-354) human Pax2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title