missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax2, Mouse anti-Human, Clone: PAX2/1104, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79742-20UG
This item is not returnable.
View return policy
Description
Pax2 Monoclonal specifically detects Pax2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Pax2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q02962, Q02962 | |
| PAX2 | |
| Recombinant fragment (around aa 223-354) human Pax2 protein (exact sequence is proprietary) | |
| 20 μg | |
| Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
| 5076 | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| PAX2/1104 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody recognizes a protein of 42kDa, which is identified as Pax2. Pax2 has been used as a marker for the identification of renal cell carcinoma and ovarian carcinoma by immunohistochemistry. | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction