missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX1 Mouse anti-Human, Mouse, Clone: 5A10D9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PAX1 |
|---|---|
| Clone | 5A10D9 |
| Concentration | 1.6 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16827235
|
Proteintech
60217-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16817235
|
Proteintech
60217-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to PAX1.
The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.Specifications
| PAX1 | |
| 1.6 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HUP48, paired box 1, Paired box protein Pax 1, PAX1 | |
| Pax1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 5A10D9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P09084, P15863 | |
| 18503, 5075 | |
| Peptide | |
| Primary | |
| -20°C | |
| PAX1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title