missing translation for 'onlineSavingsMsg'
Learn More

PAX1 Mouse anti-Human, Mouse, Clone: 5A10D9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60217-1-IG-150UL

 View more versions of this product

Product Code. 16817235

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to PAX1.

The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.
Specifications

Specifications

PAX1
Monoclonal
1.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09084, P15863
Pax1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG2b
Western Blot
5A10D9
Unconjugated
PAX1
HUP48, paired box 1, Paired box protein Pax 1, PAX1
Mouse
Protein A
RUO
18503, 5075
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.