missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX1 Mouse anti-Human, Mouse, Clone: 5A10D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60217-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PAX1.
The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.Specifications
| PAX1 | |
| Monoclonal | |
| 1.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09084, P15863 | |
| Pax1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 5A10D9 | |
| Unconjugated | |
| PAX1 | |
| HUP48, paired box 1, Paired box protein Pax 1, PAX1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 18503, 5075 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction