missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS GHSR Bioassay Kit
Description
The PathHunter™ U2OS GHSR Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay asseses ligand (e.g. Ghrelin) based on activation of GHSR GPCR activity via detection of β-Arrestin recruitment. The included cell line overexpresses ProLink-tagged GHSR and EA-tagged . Activation of GHSR stimulates the recruitment of and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | β-Arrestin recruitment |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Cell Line | U2OS |
| Target | GHSR |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?