missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS FGFR4-β-Klotho Bioassay Kit
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ FGFR4-β-Klotho Bioassay kit provides an easy-to-use cell based assay to determine FGF-19 potency and detect neutralizing antibodies. This bioassay assesses FGF19-induced activation of FGFR4-β-Klotho RTK receptors via detection of SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Dimerization |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | FGFR4-β-Klotho |
| Cell Line | U2OS |
| Target | FGFR4-β-Klotho |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction