missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ IL17RA/IL17RC Dimerization Bioassay Kit
Description
The PathHunter™ IL17RA/IL17RC Dimerization Bioassay kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This bioassay assesses ligand (e.g. IL-17) induced dimerization of IL17RA/IL17RC receptors. This product uses EFC technology. The cell line included overexpresses IL17RC target protein fused to PK alongside IL17RA target protein fused to EA. Upon ligand-induced dimerization of IL17RA/IL17RC , detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Dimerization |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | IL17RA/IL17RC |
| Cell Line | U2OS |
| Target | IL17RA/IL17RC |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?