missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ Growth Hormone Bioassay Kit
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ Growth Hormone Bioassay kit provides an easy-to-use cell based assay to determine Human Growth Hormone potency and detect neutralizing antibodies. This bioassay assesses human growth hormone-induced activation of GHR cytokine receptors via detection of SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged GHR target protein and EA-tagged SH2 domain protein. Upon activation of the receptor by its ligand or an agonist antibody, recruitment of SH2 domain protein to the phosphorylated receptor tail leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | β-Arrestin recruitment |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | GHR |
| Cell Line | U2OS |
| Target | GHR |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction