missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress PTGFR Total GPCR Internalization Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ PTGFR Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. prostaglandin F2) based internalization of PTGFR via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged PTGFR and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the PTGFR receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Total Internalization |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | (+/-)-Cloprostenol sodium salt hydrate |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | PTGFR |
| Cell Line | U2OS |
| Target | PTGFR |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction